E-4031 Dihydrochloride ≥98%
Supplier: Enzo Life Sciences
BML-KC158-0005
BML-KC158-0025
89160-234EA
368.14
CAD
89160-234
89160-236
E-4031 Dihydrochloride ≥98%
E-4031 Dihydrochloride
K⁺ channel blocker.
- Formulation: White solid.
Potent and selective blocker of the inactivating inward rectifying potassium current IKR (HERG channel). Class III antiarrythmic agent with differential atrial vs ventricular activity.
Hygroscopic and soluble in water (100 mg/mL) or DMSO.
Formula:
C₂₁H₂₇N₃O₃S·2HCl·2H₂O MW: 510.5 g/mol Storage Temperature: Ambient |
CAS Number:
113558-89-7 |
|
Specification Test Results
Purity | ≥98% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...