Penitrem A ≥95% (by HPLC, TLC)

Supplier: Enzo Life Sciences
Danger

BML-KC157-0001 BML-KC157-0005
89160-230EA 116.92 CAD
89160-230 89160-232
Penitrem A ≥95% (by HPLC, TLC)

K⁺ channel blocker.


  • Formulation: White solid.


Fungal mycotoxin. Selective, irreversible blocker of the smooth muscle high conductance Ca²⁺-activated K⁺ (maxi-K) channel (100% block at 10 nM). Increases spontaneous release of GABA and aspartate from cerebrocortical synaptosomes in rat neuromuscular junction.


Soluble in organic solvents such as methanol, acetone or DMSO.


Caution: Stable for at least 1 year after receipt when stored, as supplied, at -20 °C. Stock solutions are stable for up to 3 months at -20 °C.

Formula: C₃₇H₄₄ClNO₆
CAS Number: 12627-35-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d3ae7822ca45998c9ef16b893efab3c2ef5b021&vid=20ab5fb0125c5baa2f6eff1c2a8df8c8be154f8d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥95% (HPLC, TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR