(S)-Mephenytoin ≥98% (by TLC)

Supplier: Enzo Life Sciences
Warning

BML-J120-0005 BML-J120-0025
89160-172EA 170.64 CAD
89160-172 89160-174
(S)-Mephenytoin ≥98% (by TLC)
(S)-Mephenytoin

Cytochrome P450 substrate.


  • Formulation: White solid.


Substrate for determination of cytochrome P-450 polymorphism. Anticonvulsant.


Soluble in DMSO or ethanol.

Formula: C₁₂H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00270025
CAS Number: 70989-04-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90be63fb993e71ee655e11959976de69ec38d8ed&vid=a6c6bc4f234423b4dac7d68f1637900c37ca7b47&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR