Imipramine-N-oxide ≥98%, Ultrapure

Supplier: Enzo Life Sciences
Danger

BML-J116-0005 BML-J116-0025
89160-168EA 188.02 CAD
89160-168 89160-170
Imipramine-N-oxide ≥98%, Ultrapure
Imipramine-N-oxide

Imipramine metabolite produced via flavin-containing monooxygenase-mediated oxidation.


  • Formulation: White to off-white solid


Soluble in DMSO (30 mg/mL), ethanol (20 mg/mL) or water (2 mg/mL warm).


Caution: Store, as supplied, at room temperature for up to 1 year. Store solutions at -20 °C for up to 3 months.

Formula: C₁₉H₂₄N₂O
MW: 296.4 g/mol
Storage Temperature: Ambient
CAS Number: 6829-98-7
UN: 2811
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c43c8c6868e3f0a66a51d4000a13c91b9409d0e&vid=61bdf68b974a39f123ece39904c52998ce1df422&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR