Despropionylfentanyl ≥99%

Supplier: Enzo Life Sciences
Warning

BML-J110-0001 BML-J110-0010
89160-160EA 146.94 CAD
89160-160 89160-162
Despropionylfentanyl ≥99%
Despropionylfentanyl

A major urinary metabolite of fentanyl.


  • Formulation: Off-white solid.


Soluble in 100% ethanol.

Formula: C₁₉H₂₄N₂
Storage Temperature: Ambient
CAS Number: 21409-26-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fff514de79f2fc79ea268ac525113282ed3650ff&vid=a337744a1934621d6b2fe5295356daf858fea8ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Source Synthetic
Purity ≥99% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR