Mycophenolate mofetil ≥98% (by HPLC), white to off-white solid

Supplier: Enzo Life Sciences
Warning

BML-IM101-0020 BML-IM101-0100
89160-152EA 139.04 CAD
89160-152 89160-154
Mycophenolate mofetil ≥98% (by HPLC), white to off-white solid
Mycophenolate mofetil

Immunosuppressant. Isolated from Penicillium brevicompactum.


  • Formulation: White to off-white solid.


Mycophenolate mofetil (cellcept) is an immunosuppressant prodrug of mycophenolic acid, a fermentation product of Penicillium brevicompactum. Immunosuppression occurs via inhibition of type II inosine-5’-monophosphate dehydrogenase, depleting guanosine preferentially in T and B lymphocytes. Mycophenolic acid displays anti-inflammatory activity by preventing NO production specifically from iNOS and by blocking glycosylation and expression of adhesion molecules and therefore recruitment of lymphocytes and monocytes to sites of inflammation.


Soluble in DMSO (25 mg/mL) or 100% ethanol (25 mg/mL, warm).

Formula: C₂₃H₃₁NO₇
MDL Number: MFCD00867568
CAS Number: 128794-94-5
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=710a733e215b5b423b89eb1aa5111671e0a8138a&vid=5ad4befb32a61bd07a760f71ee0a87dda15d96d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Appearance White to off-white solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR