13(S)-Hydroperoxyocta-deca-9Z,11E-dienoic acid ≥98%
Supplier: Enzo Life Sciences
BML-HP013-0050
BML-HP013-1000
89160-098EA
165.9
CAD
89160-098
89160-100
13(S)-Hydroperoxyocta-deca-9Z,11E-dienoic acid ≥98%
13(S)-Hydroperoxyocta-deca-9Z,11E-dienoic acid
Suppresses activation of platelets. Induces endothelial injury. Constricts isolated coronary arteries. Inhibits prostacyclin synthase.
- Inhibits prostacyclin synthase
Suppresses activation of platelets. Induces endothelial injury. Constricts isolated coronary arteries. Inhibits prostacyclin synthase.
Formula:
C₁₈H₃₂O₄ Storage Temperature: Ultra-Low Temperature Freezer (–80°C) |
CAS Number:
33964-75-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...