4-Hydroxynonenal ≥97%
Supplier: Enzo Life Sciences
Warning
|
BML-HA004-0001
BML-HA004-0010
89160-046EA
162.74
CAD
89160-046
89160-048
4-Hydroxynonenal ≥97%
4-Hydroxynonenal
Lipid peroxidation.
- Formulation: Colorless oil.
Cytotoxic product of microsomal lipid peroxidation induced by xenobiotics. Reported to be a specific chemical indicator of lipid peroxidation in rat liver microsomes and in myocardial reperfusion injury. Covalently reacts with LDL and HDL during oxidative modification. Modifies proteins by covalent reaction with histidine and lysine residues. Activates TRPA1.
Soluble in 100% ethanol (25 mg/mL).
Formula:
C₉H₁₆O₂ Storage Temperature: Ultra-Low Temperature Freezer (–80°C) |
MDL Number:
MFCD00211020 CAS Number: 75899-68-2 |
|
Specification Test Results
Purity | ≥97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...