BML-278 ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-GR359-0005 BML-GR359-0025
89158-996EA 158 CAD
89158-996 89158-998
BML-278 ≥98% (by TLC)
BML-278

A SIRT1 activator.


  • Formulation: Off-white solid.


Cell-permeable. A structurally novel activator of SIRT1 (EC₅₀ = 1 µM). Displays selectivity over SIRT2 and 3 (EC₅₀ = 25 and 50 µM respectively).


Soluble in DMSO (~5 mg/mL).

Formula: C₂₄H₂₅NO₄
Storage Temperature: Ambient
CAS Number: 15301-69-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d663d03aaef27496f186307811ce3ea5327d5c1&vid=b0e7c17daee2247bd45f84a362a88f7d2d7436e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% by TLC

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR