BML-266 ≥95% (by TLC)
Supplier: Enzo Life Sciences
Synonyms:
JFD00244, 1,4-Di[(4-hydroxyphenethyl)amino]-9,10-dihydroanthracene-9,10-dione
BML-GR346-0010
BML-GR346-0050
89158-982EA
237
CAD
89158-982
89158-984
BML-266 ≥95% (by TLC)
BML-266
Structurally novel SIRT2 inhibitor (IC50=56.7µM).
- SIRT inhibitor
Dark purple solid.
Soluble in DMSO (25mg/ml).
Formula:
C₃₀H₂₆N₂O₄ MW: 478.5 g/mol Storage Temperature: Freezer |
CAS Number:
96969-83-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...