Garcinol ≥95% (by TLC)
Supplier: Enzo Life Sciences
BML-GR343-0010 BML-GR343-0050
89158-978EA 237 CAD
89158-978 89158-980
Garcinol ≥95% (by TLC)
Garcinol
HAT inhibitor
Garcinol is a polyisoprenylated benzophenone derivative isolated from Garcinia indica. It is a potent inhibitor of histone acetyltransferases (HATs) p300 (IC50=7µM) and PCAF (IC50=5µM) both in vitro and in vivo. HAT activity-dependent chromatin transcription was inhibited by garcinol, but it had no effect on naked DNA transcription. It induces apoptosis in HeLa cells (100µM) where it down regulates global gene expression. Acetylation of histone H4 by p300 was more sensitive to inhibition by garcinol compared with that of H3. Histone H4 acetylation was completely inhibited at 1 µM while acetylation of H3 was not abolished at 20 µM.
CAS Number: 78824-30-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfdf277e82839de4b32a76336b01929887e432e3&vid=0d8c3228c4c0390e0475805edd086128c2484fcf&prid=web&clid=SAPDAM