Dimethyl-bisphenol A ≥98% (by TLC)
Supplier: Enzo Life Sciences
Synonyms:
4,4'-(Propane-2,2-diyl)bis(methoxybenzene)
BML-GR339-0010
BML-GR339-0050
89158-966EA
180.12
CAD
89158-966
89158-968
Dimethyl-bisphenol A ≥98% (by TLC)
Dimethyl-bisphenol A
A HIF-1α degradation promotor.
- HIF-1α inhibitor
White solid.
Soluble in DMSO (>25 mg/ml) or ethanol (>25 mg/ml)
Formula:
C₁₇H₂₀O₂ MW: 256.3 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00984727 CAS Number: 1568-83-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...