BML-210 ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: CAY10433, N-(2-aminophenyl)-N'-phenyl-octanediamide, N1-(2-Aminophenyl)-N8-phenyloctanediamide

BML-GR330-0001 BML-GR330-0005
89158-940EA 172.22 CAD
89158-940 89158-942
BML-210 ≥98% (by TLC)
BML-210

HDAC inhibitor developed at Enzo Life Sciences. Inhibits HDAC activity in HeLa nuclear extracts (IC50=5-10µM).


  • HDAC inhibitor


Tan solid.


Soluble in DMSO (25mg/ml) or 100% ethanol (10mg/ml, warm)

Formula: C₂₀H₂₅N₃O₂
Storage Temperature: Freezer
MDL Number: MFCD08062139
CAS Number: 537034-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f4e13ae38c8b295361e899011e590c3638a9940&vid=68f17a50e559d4c67e71d6d33fe5a1a6d2f6cd60&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR