Nullscript ≥97% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: 4-(1,3-Dioxo-1H,3H-benzo[de]isoquinolin-2-yl)-N-hydroxybutanamide

BML-GR327-0001 BML-GR327-0005
89158-936EA 143.78 CAD
89158-936 89158-938
Nullscript ≥97% (by TLC)
Nullscript

HDAC inhibitor


Negative control compound for the histone deacetylase (HDAC) inhibitor scriptaid (Prod. No. BML-GR326).

Formula: C₁₆H₁₄N₂O₄
MW: 298.30 g/mol
MDL Number: MFCD00405081
CAS Number: 300816-11-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b2118736bcf936201c23a0f64172de0fed869a6&vid=1b390781868ed27c215e94ee42dddbc17ae44567&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR