scriptaid ≥97% (by TLC)

Supplier: Enzo Life Sciences

BML-GR326-0001 BML-GR326-0005
89158-932EA 143.78 CAD
89158-932 89158-934
scriptaid ≥97% (by TLC)
scriptaid

HDAC inhibitor


Inhibitor of histone deacetylase (HDAC) (HDAC1 and 3: IC50~0.6µM; HDAC8: IC50~1µM) with lower toxicity than trichostatin A (Prod. No. BML-GR309). Inhibits tumor growth in vitro and in vivo and, in conjunction with 5-aza-2’-deoxycytidine (Prod. No. ALX-480-096), enhances expression of estrogen receptor α. Optimal concentration 2-2.5μg/ml (6-8μM). A negative control is available (Prod. No. BML-GR327).

Formula: C₁₈H₁₈N₂O₄
MDL Number: MFCD00386477
CAS Number: 287383-59-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be932dbb3fc124e96e8e126524c22724505e680f&vid=ac69561e0f0f395777ef7d0145caa1ef2cd19d51&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR