DIF-1 ≥98% (by HPLC), Ultrapure
Supplier: Enzo Life Sciences
BML-GR324-0100
BML-GR324-0500
89158-924EA
140.62
CAD
89158-924
89158-926
DIF-1 ≥98% (by HPLC), Ultrapure
DIF-1
Induces cell differentiation.
- Formulation: Pale yellow solid.
Supresses cell growth and promotes retinoic acid-induced cell differentiation in HL-60 cells. It is a known inhibitor of Wnt/β-Catenin signaling pathway.
Soluble in ethanol.
Formula:
C₁₃H₁₆Cl₂O₄ MW: 307.2 g/mol Storage Temperature: Freezer |
CAS Number:
111050-72-7 |
|
Specification Test Results
Purity | ≥98% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...