Monastrol ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-GR322-0005 BML-GR322-0025
89158-916EA 287.56 CAD
89158-916 89158-918
Monastrol ≥98% (by TLC)
Monastrol

Cell permeable non-tubulin-interacting mitosis inhibitor. Blocks mitosis (IC50=14µM) by binding to the mitotic kinesin Eg5.


  • Kinesin inhibitor


White to light yellow solid.


Soluble in DMSO (20mg/ml) or 100% ethanol (4mg/ml).

Formula: C₁₄H₁₆N₂O₃S
Melting Pt: 184…186 °C
Storage Temperature: Freezer
MDL Number: MFCD00813077
CAS Number: 329689-23-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4859fddc7985a67ca50ceaea49b9a384cfcf104d&vid=86466f37e275695802e2d24d97ff9a88e30ffc6b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR