(±)-10-Hydroxycamptothecin ≥97%

Supplier: Enzo Life Sciences

Synonyms: 1-HCPT, 10-Hydroxycamptothecin, 4-Ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, NSC107124

BML-GR316-0020 BML-GR316-0100
89158-898EA 172.22 CAD
89158-898 89158-900
(±)-10-Hydroxycamptothecin ≥97%
(±)-10-Hydroxycamptothecin

A DNA topoisomerase I inhibitor that induces differentiation in HL-60 cells. It induces apoptosis in human breast cancer cell lines MCF-7 and MDA-MB-468.


  • Topoisomerase inhibitor


Pale yellow solid.


Soluble in DMSO.

Formula: C₂₀H₁₆N₂O₅
MW: 364.35 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09027971
CAS Number: 64439-81-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80a3bacbeb95cd3f8e6cbaae980b109b3f52d716&vid=170ff37013d543efda901cc7a36fcc7f77503c63&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR