Ellipticine ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: 5,11-Dimethyl-6H-pyrido[4,3-b]carbazole

BML-GR315-0010 BML-GR315-0050
89158-894EA 211.72 CAD
89158-894 89158-896
Ellipticine ≥98% (by TLC)
Ellipticine

A DNA-intercalating antitumor agent. Inhibits DNA topoisomerase II. Inhibits the phosphorylation of p53 in Lewis lung carcinoma cells and induces apoptosis. Inhibits cytochrome P-450-1A1.


  • Topoisomerase inhibitor


Bright yellow solid.


Soluble in DMSO, DMF or ethanol.

Formula: C₁₇H₁₄N₂
Storage Temperature: Freezer
MDL Number: MFCD00010524
CAS Number: 519-23-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1952e861a593e8439f480a4cd6408388c22871e0&vid=7001ffa95da6c50564c094994ef917d10dfb1b7e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR