Lycorine HCl ≥97%

Supplier: Enzo Life Sciences

BML-GR313-0010 BML-GR313-0050
89158-886EA 167.48 CAD
89158-886 89158-888
Lycorine HCl ≥97%
Lycorine HCl

Lycorine is a eukaryotic termination inhibitor.


  • Protein synthesis inhibitor


White to off-white powder.


Soluble in water (>15mg/ml).

Formula: C₁₆H₁₇NO₄·HCl
Storage Temperature: Freezer
MDL Number: MFCD00243111
CAS Number: 2188-68-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=112fe242c9c173f1299f33acf3ddffd3994febb4&vid=a5c4d1238478138c3c7877549f1cb46954c16f3a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR