Puromycin dihydrochloride ≥98% (by HPLC)
Supplier: Enzo Life Sciences
BML-GR312-0050
BML-GR312-0250
89158-882EA
121.66
CAD
89158-882
89158-884
Puromycin dihydrochloride ≥98% (by HPLC)
Puromycin dihydrochloride
Inhibits protein synthesis
- Appearance: White to off-white solid
- Soluble in water (50 mg/mL)
Inhibits translation by causing premature release of nascent polypeptide. Induces apoptosis.
Formula:
C₂₂H₂₉N₇O₅·2HCl Storage Temperature: Freezer |
MDL Number:
MFCD00012691 CAS Number: 58-58-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...