Etoposide ≥97% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: (5R,5aR,8aR,9S)-9-(((4aR,6R,7R,8R,8aS)-7,8-Dihydroxy-2-methylhexahydropyrano[3,2-d][1,3]dioxin-6-yl)oxy)-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one, 4'-Demethylepipodophyllotoxin 9-(4,6-O-Ethylidene-beta-D-glucopyranoside)

BML-GR307-0100 BML-GR307-0500
89158-866EA 184.86 CAD
89158-866 89158-868
Etoposide ≥97% (by HPLC)
Etoposide

Topoisomerase inhibitor


Antitumor reagent. Topoisomerase II inhibitor. Induces apoptosis by FasL. Activates p53 resulting in upregulated expression of TRAIL-R2 (DR5) and Bak to overcome TRAIL resistance in Bax-deficient human colon carcinoma cancer cells.


Semisynthetic derivative of podophyllotoxin.


p53 activator.

Formula: C₂₉H₃₂O₁₃
MW: 588.56 g/mol
MDL Number: MFCD00869325
CAS Number: 33419-42-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee83d78623b759133b10fb94a5ff19a5c40f9371&vid=762d7a838872b1d46f7d48d5deeaa8ae212a2e41&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR