SR-12813 ≥98%

Supplier: Enzo Life Sciences

BML-GR242-0010 BML-GR242-0050
89158-834EA 180.12 CAD
89158-834 89158-836
SR-12813 ≥98%
SR-12813

PXR activator


Potent and selective PXR agonist. (EC50=200 nM for human PXR). A weak activator of rat and mouse PXR. Inhibits cholesterol biosynthesis in a number of species.

Formula: C₂₄H₄₂O₇P₂
MDL Number: MFCD00888473
CAS Number: 126411-39-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=676be6f50009ee6bb0368c2d3af1ab0fcba57d37&vid=ebec33e5ed84efeceb452906af8b4bd678466212&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR