GW-9662 ≥98%

Supplier: Enzo Life Sciences
Warning

Synonyms: GW9662, 2-Chloro-5-nitro-N-phenylbenzamide

BML-GR234-0010 BML-GR234-0050
89158-818EA 175.38 CAD
89158-818 89158-820
GW-9662 ≥98%
GW-9662

PPAR ligand.


  • Formulation: White to off-white solid.


Potent, selective and irreversible PPARγ antagonist. Blocks the PPARγ-induced differentiation of monocytes to osteoclasts by >90% at a dose of 0.1 µM. Can be used to antagonise the effects of PPARγ agonists in a variety of cell types.


Soluble in DMSO (50 mg/mL), 100% ethanol (10 mg/mL) or dimethyl formamide. Insoluble in water.

Formula: C₁₃H₉N₂O₃Cl
Melting Pt: 155…157 °C
MDL Number: MFCD01215270
CAS Number: 22978-25-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e921714d1c9b480af8862b60e9530751471116d&vid=930ede030b5767df965756e8f6c6b3f9981d185b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR