24(S),25-Epoxycholesterol ≥98% (by TLC)

Supplier: Enzo Life Sciences


Packaged under inert gas.

BML-GR231-0001 BML-GR231-0005
89158-810EA 436.08 CAD
89158-810 89158-812
24(S),25-Epoxycholesterol ≥98% (by TLC)
24(S),25-Epoxycholesterol

LXR ligand.


  • Formulation: White solid.


Endogenous agonist for the nuclear receptor LXR.


Soluble in ethanol (10 mg/mL) or DMSO (10 mg/mL).


Caution: Should be used immediately following dissolution. Store with desiccant.

Formula: C₂₇H₄₄O₂
Melting Pt: 156…159 °C
CAS Number: 77058-74-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c11fa0dcf085da13cdb6fb657808bb83018888f&vid=09cb0ef6a0e47d0914adacd42cb1207c1135fefc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (TLC)
Identity Determined by NMR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR