2-Chloro-5-nitro-N-(4-pyridyl)benzamide ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-GR212-0010 BML-GR212-0050
89158-802EA 192.76 CAD
89158-802 89158-804
2-Chloro-5-nitro-N-(4-pyridyl)benzamide ≥98% (by TLC)
2-Chloro-5-nitro-N-(4-pyridyl)benzamide

Potent and selective PPARγ antagonist. T0070907 blocks PPARγ function in both cell-based reporter gene and adipocyte differentiation assays. It inhibits growth and survival of hepatocellular carcinoma cells and squamous cell carcinoma cell lines by interfering with cell adhesion "anoikis".


  • PPAR ligand


Off-white solid.


Soluble in DMSO (>25 mg/ml).

Formula: C₁₂H₈ClN₃O₃
Storage Temperature: Ambient
MDL Number: MFCD00121849
CAS Number: 313516-66-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cb75d0447e6b82aded70f96dee6614230e1279a&vid=8498d6d44cb722f9b57f4e12c5ac229864299f4e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR