AC-55469 ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 4'-Octyl-[1,1'-biphenyl]-4-carboxylic acidAC-55649

BML-GR112-0050 BML-GR112-0010
89158-778EA 600.95 CAD
89158-778 89158-776
AC-55469 ≥98% (by HPLC)
AC-55469

AC-55649 is potent and subtype selective agonist at RARβ2 ( EC50=6.9 µM). It inhibits proliferation of MCF-7 cells and induces differentiation in a neuronal cell line.


  • Retinoic acid receptor ligand


White to gray, Crystals or crystalline powder

MW: 310.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00157040
CAS Number: 59662-49-6
https://ca.vwr.com/stibo/web/std.lang.all/39/22/45403922.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR