AM 580 ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-GR104-0005 BML-GR104-0025
89158-748EA 237 CAD
89158-748 89158-750
AM 580 ≥98% (by TLC)
AM 580

Retinoic acid analog which acts as a selective RARα agonist with no affinity for RXRs. Induces differentiation in leukemia cell lines and inhibits angiogenesis in chick embryos.


  • Retinoic acid receptor ligand


White to off-white solid.


Soluble in DMSO (25mg/ml) and ethanol (10mg/ml).

Formula: C₂₂H₂₅NO₃
Storage Temperature: Freezer
MDL Number: MFCD00673916
CAS Number: 102121-60-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbca291264cfaa35adf2204220afbb5b6c286be4&vid=c6d9ccedeac2847b0a7ee3738d0bcc622bd166b9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR