9-cis-Retinoic acid ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Danger

BML-GR101-0005 BML-GR101-0025
89158-736EA 308.1 CAD
89158-736 89158-738
9-cis-Retinoic acid ≥98% (by HPLC)
9-cis-Retinoic acid

Retinoid X receptor ligand.


  • Formulation: Light yellow to orange crystalline powder.


A high affinity ligand for the retinoid X receptor (RXR). Destabilizes VDR-RXR heterodimer-DNA complex.


Soluble in DMSO (25 mg/mL) or 100% ethanol (25 mg/mL).

Formula: C₂₀H₂₈O₂
MDL Number: MFCD00270072
CAS Number: 5300-03-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00ce44a39231d676886ade8b949ae3e44a3f6b93&vid=8b26c637c8947137019fbe2a1d51fecd636ed6b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR