Ar231453 ≥98%, yellow solid

Supplier: Enzo Life Sciences

Synonyms: N-(2-Fluoro-4-(methylsulfonyl)phenyl)-6-(4-(3-isopropyl-1,2,4-oxadiazol-5-yl)piperidin-1-yl)-5-nitropyrimidin-4-amine, N-(2-Fluoro-4-(methylsulphonyl)phenyl)-6-(4-(3-isopropyl-1,2,4-oxadiazol-5-yl)piperidin-1-yl)-5-nitropyrimidin-4-amine, AR 231453

BML-GP100-0005 BML-GP100-0025
89158-710EA 158 CAD
89158-710 89158-712
Ar231453 ≥98%, yellow solid
Ar231453

GPR119 activator.


  • Formulation: Yellow solid.


Synthetic agonist of the orphan GPR119. Although endogenous lipid amides such as oleoyl ethanolamide have been reported to be agonists at GPR119, AR231453 is the first reported high potency agonist (EC50=0.68nM). The compound displayed in vivo activity in rodents and was active in an oral glucose tolerance test in mice following oral administration. AR231453 stimulated the release of GLP-1 via GPR119 receptors in mouse models. It was shown to be a highly selective ligand with no off-target activity across a 76 receptor and enzyme profiling panel (CEREP) and no activity against 140 known and orphan GPCRs in melanophore dispersion assays.


Soluble in DMSO (>25 mg/mL).

Formula: C₂₁H₂₄FN₇O₅S
Storage Temperature: Freezer
MDL Number: MFCD14706714
CAS Number: 733750-99-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d2db09bd18661e398f0381bc7bdc38bfc70cda3&vid=9afdcb41001a30f0fa7d109d644bbfb8267e992f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Identity Determined by NMR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR