Anti-GAD1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
GC3008-0025 GC3008-0100
89158-702EA 222.78 CAD
89158-702 89158-704
Anti-GAD1 Rabbit Polyclonal Antibody
Antibodies
Glutamic acid decarboxylase (GAD; E.C. 4.1.1.15) is the enzyme responsible for the conversion of glutamic acid to γ-aminobutyric acid (GABA) - the major inhibitory transmitter in higher brain regions. Two molecular forms of GAD (GAD65 and GAD67) are known from rat, cat, pig and man, and both forms are expressed in the CNS and in pancreatic islet endocrine cells. A recent Western blotting study has identified that the isoforms are regionally distributed in the brains of rats and mice. In rat pancreas, GAD65 and GAD67 appear to be differentially localised, GAD65 primarily in insulin-containing (β) cells and GAD67 in glucagon-containing (A) cells. Evidence from knock-out mice suggests that GABA generated by pancreatic GAD isoforms is not critically involved in normal islet cell development and morphology. However, β-cell-specific GAD expression has been shown to be required for the development of autoimmune diabetes in non-obese diabetic (NOD) mice.

  • For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

Recognizes human, mouse, rat and cat GAD. Detects two isoforms of ~65kDa and ~67 kDa by Western Blot.

Type: Primary
Antigen: GLUTAMATE DECARBOXYLASE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR