Simvastatin ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-G244-0050 BML-G244-0250
89158-612EA 267.02 CAD
89158-612 89158-614
Simvastatin ≥98% (by TLC)
Simvastatin

HMG-CoA reductase inhibitor


HMG-CoA reductase inhibitor. Synthetic analog of Lovastatin (Prod. No. BML-G226). Inhibits the biosynthesis of isoprenoids and prevents small G protein activation. Displays lipid lowering and antihypertensive effects. Displays antiinflammatory, immunomodulatory effects as well as protective effects in experimental traumatic brain injury. Activates Rac1-dependent signalling pathway.

Formula: C25H38O5
MDL Number: MFCD00072007
CAS Number: 79902-63-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9623c52f40d73b09695fcf40beecb470b45d4337&vid=f9d3d6fedc4cb8adb520fb175b92ad84c306b6ef&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR