Geranylgeraniol ≥95% (by TLC), colourless oil

Supplier: Enzo Life Sciences
Warning

BML-G243-0020 BML-G243-0100
89158-608EA 183.28 CAD
89158-608 89158-610
Geranylgeraniol ≥95% (by TLC), colourless oil
Geranylgeraniol

Geranylgeranylpyrophosphate precursor and apoptosis inducer.


  • Formulation: Colorless Oil.


Induces apoptosis in HL-60 cells.


Soluble in chloroform or ethyl acetate.

Formula: C₂₀H₃₄O
Storage Temperature: Freezer
MDL Number: MFCD00129083
CAS Number: 24034-73-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbe048b645fdb33fb0def17564b9a83cba1ced3f&vid=6f57638e835f319993613ed0abface1102378b32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥95% (TLC)
Identity Determined by H-NMR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR