B581 ≥96%

Supplier: Enzo Life Sciences

BML-G235-0001 BML-G235-0005
89158-592EA 331.8 CAD
89158-592 89158-594
B581 ≥96%
B581

Farnesyltransferase inhibitor


A potent and selective, cell-permeable farnesyl transferase inhibitor IC50=21 nM, 30-fold selective over geranylgeranyl transferase.

Formula: C₂₂H₃₈N₄O₃S₂
MDL Number: MFCD18910841
CAS Number: 149759-96-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5cef061310b7dac5bbeccb5ced37b03b8f16c75&vid=f80710c6888ef5dc094df34a6e4c4a8cb49f5582&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR