12-Methoxydodecanoic acid ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-G234-0010 BML-G234-0050
89158-588EA 140.62 CAD
89158-588 89158-590
12-Methoxydodecanoic acid ≥98% (by TLC)
12-Methoxydodecanoic acid

A myristate analog which has similar chain length but reduced hydrophobicity compared to myristate. Inhibits replication of HIV-1. Its specific mechanism of action is not clear but it is believed to either inhibit the activity of N-myristoyltransferase or modulate the function of target acylated proteins.


  • Myristate analog


White to off-white solid.


Soluble in ethanol (25 mg/mL) and DMSO (25 mg/mL).

Formula: C₁₃H₂₆O₃
MW: 230.348 g/mol
MDL Number: MFCD00899847
CAS Number: 92169-28-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65c59413cc056d8ca9f07c472391d2a6a623e6f8&vid=0c76b60f74f4ae1e83888ec462f877140ac20031&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR