Mevastatin ≥95%

Supplier: Enzo Life Sciences

Synonyms: (1S,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-7-methyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (S)-2-methylbutanoate, Compactin

BML-G233-0010 BML-G233-0050
89158-584EA 153.26 CAD
89158-584 89158-586
Mevastatin ≥95%
Mevastatin

HMG-CoA reductase inhibitor


Mevastatin inhibits isoprenoid biosynthesis by inhibition of HMG-CoA reductase (Ki for acid form is 1 nM) and therefore blocks protein isoprenylation and reduces plasma cholesterol levels in humans. It causes cells to arrest early in the G1 phase. Mevastatin is a close structural analog of lovastatin and both agents have the same biochemical and pharmacological activities. Mevastatin is inactive in cell-free assays. In cells however, it is hydrolyzed to the active free acid form by intracellular esterases.

Formula: C₂₃H₃₄O₅
MW: 390.52 g/mol
MDL Number: MFCD05662341
CAS Number: 73573-88-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a078f33ec862296b038eb7e7b792b036e75ea551&vid=2ed5aca0906fab3312cc574166f4280c15d93ab3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR