Chaetomellic acid A ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-G229-0005 BML-G229-0025
89158-568EA 131.14 CAD
89158-568 89158-570
Chaetomellic acid A ≥98% (by TLC)
Chaetomellic acid A

Potent inhibitor of farnesyltransferase in isolated enzyme assays (IC50=55nM) but is inactive in whole cells. Inhibition of farnesyltransferase is selective over geranylgeranyltransferase I and II (IC50=92µM and 34µM respectively).


  • Farnesyltransferase inhibitor


White to off-white solid.


Soluble in warm water (2mg/ml, slightly cloudy).

Formula: C₁₉H₃₂O₄·2Na
Melting Pt: >315 °C
Storage Temperature: Freezer
CAS Number: 148796-51-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77c7a96ffe7c7c1c25a430bd5769c9231d87177a&vid=4ffbb4d009e0a0c864a38f354128e27b82850cef&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR