Lovastatin ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-G226-0010 BML-G226-0050
89158-562EA 105.86 CAD
89158-562 89158-564
Lovastatin ≥98% (by TLC)
Lovastatin

HMG-CoA reductase inhibitor


Inhibits cholesterol/isoprenoid biosynthesis by inhibition of HMG-CoA reductase (Ki for acid form is 1nM) and blocks protein isoprenylation and reduces plasma cholesterol levels in humans. Causes cells to arrest early in the G1 phase. Induces apoptosis in cancer cells.

Formula: C₂₄H₃₆O₅
MDL Number: MFCD00072164
CAS Number: 75330-75-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ac7c44bb3bfcacb1f1152a60c37a2bd8c825610&vid=02e5429a451616e60503ccb6dd01b8eaa8d9641b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR