Farnesylthioacetic acid ≥98%

Supplier: Enzo Life Sciences

BML-G222-0005 BML-G222-0025
89158-552EA 158 CAD
89158-552 89158-554
Farnesylthioacetic acid ≥98%
Farnesylthioacetic acid

Methyltransferase inhibitor


FTA potently and specifically inhibits methyl esterification of farnesylated proteins such as the γ subunit of transducin (70% at 10µM). It does not act as a substrate for methyl transferase.

Formula: C₁₇H₂₈O₂S
MDL Number: MFCD00216146
CAS Number: 135784-48-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a88bd2e1dbef027b42484c58c1f2ff00441c2a8&vid=401e20a118c42d943a16a3274d37a96579a21a8a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR