N-Acetyl-S-geranylgeranyl-L-cysteine ≥98% (by TLC)
Supplier: Enzo Life Sciences
Synonyms:
AGGC
BML-G220-0005
BML-G220-0025
89158-544EA
183.28
CAD
89158-544
89158-546
N-Acetyl-S-geranylgeranyl-L-cysteine ≥98% (by TLC)
N-Acetyl-S-geranylgeranyl-L-cysteine
Blocks methyl esterification of geranylgeranylated proteins
AGGC specifically blocks methyl esterification of geranylgeranylated proteins such as Rab proteins. AGGC blocks receptor-mediated signal transduction in human neutrophils (IC50=4 µM). Induces insulin release from HIT-T15 cells by mimicking the action of Rab3A and inhibits beta2-integrin-induced actin polymerization (IC50~45 nM) in neutrophils.
CAS Number:
139332-94-8 |