S-Farnesyl-L-cysteine methyl ester (synthetic) ≥98% (by TLC), colourless oil

Supplier: Enzo Life Sciences

BML-G201-0005 BML-G201-0025
89158-536EA 178.54 CAD
89158-536 89158-538
S-Farnesyl-L-cysteine methyl ester (synthetic) ≥98% (by TLC), colourless oil
S-Farnesyl-L-cysteine methyl ester

Multidrug resistance transporter activator.


  • Formulation: Colorless oil.


Stimulates the multidrug resistance transporter ATPase activity (4-5-fold at 10-20 µM) and competes for drug binding.


Soluble in DMSO (25 mg/mL) and ethanol (25 mg/mL).

Formula: C₁₉H₃₃NO₂S
Storage Temperature: Freezer
CAS Number: 125741-64-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24a99096675aaced0509062c726e837ff148ad1e&vid=7e1d54cd8d94d2c4d66b1e4ba4464e1604f0ee95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (TLC)
Source Synthetic


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR