Idebenone ≥98%

Supplier: Enzo Life Sciences

Synonyms: 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methyl-1,4-benzoquinone, 2-(10-Hydroxydecyl)-5,6-dimethoxy-3-methylcyclohexa-2,5-diene-1,4-dione

BML-FR114-0020 BML-FR114-0100
89158-498EA 172.22 CAD
89158-498 89158-500
Idebenone ≥98%
Idebenone

Quinone antioxidant.


  • Formulation: Orange solid.


Idebenone is a quinone antioxidant chemically related to ubiquinone. It is a free radical scavenger that protects mitochondrial membranes from lipid peroxidation. It improves overall brain function in various animal models and protective effect on astrocytes is due in part via increased NGF production.

Formula: C₁₉H₃₀O₅
Storage Temperature: Ambient
MDL Number: MFCD00274552
CAS Number: 58186-27-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=780ceb87c69d3485f05308333972e2c6cd06408a&vid=94559cf65f51c61e5d28b606149c848ea97af91b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (TLC)
Appearance Orange solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR