L-Ergothioneine ≥98% (by TLC), white solid

Supplier: Enzo Life Sciences

Synonyms: (S)-3-(2-Thioxo-2,3-dihydro-1H-imidazol-4-yl)-2-(trimethylammonio)propanoate, Ergothioneine

BML-FR111-0010 BML-FR111-0050
89158-488EA 237 CAD
89158-488 89158-490
L-Ergothioneine ≥98% (by TLC), white solid
L-Ergothioneine

Antioxidant.


  • Water soluble


Naturally occuring ubiquitous antioxidant which is found in many cells at concentrations up to 2 mM. Peroxynitrate ONOO- scavenger. Protects human neuronal hybridoma cell line (N-18-RE-105) from ONOO--induced oxidative damage.

Formula: C₉H₁₅N₃O₂S
MW: 229.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00167474
CAS Number: 497-30-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=927ad62b3350ab9f4e97037cff5ad5f71be0d393&vid=5795dc3afb92ab5292dabb56c6d614c07b5027d6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR