3-Methyl-1-phenyl-2-pyrazolin-5-one ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Warning

BML-FR103-0050 BML-FR103-0250
89158-474EA 105.86 CAD
89158-474 89158-476
3-Methyl-1-phenyl-2-pyrazolin-5-one ≥98% (by HPLC)
3-Methyl-1-phenyl-2-pyrazolin-5-one

A free radical scavenger and antioxidant. Displays beneficial effects in a rat model of ischemic brain damage.


  • Formulation: White to beige to yellow powder.


Soluble in DMSO.

Formula: C₁₀H₁₀N₂O
MDL Number: MFCD00003138
CAS Number: 89-25-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6f0ac378de9f4d1748ec0c286568f5c7d13be14&vid=63fb9373f5df4649427deb61fe6e74e8b589cc57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Appearance White to beige to yellow powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR