Linoleamide ≥98% (by TLC), white solid

Supplier: Enzo Life Sciences

Synonyms: (9Z,12Z)-9,12-Octadecadienamide, Linoleic acid amide, (9Z,12Z)-octadeca-9,12-dienamide

BML-FA023-0010 BML-FA023-0050
89158-378EA 110.6 CAD
89158-378 89158-380
Linoleamide ≥98% (by TLC), white solid
Linoleamide

Unsaturated analog of endogenous sleep inducing lipid.


  • Formulation: White solid.


Soluble in 100% ethanol or in DMSO.

Formula: C₁₈H₃₃NO
MW: 279.460 g/mol
MDL Number: MFCD00798239
CAS Number: 3999-01-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f845a75711e1331b25bf57ccf753cfb9c06637e&vid=10c0afa086bbcef33061ea3c0d6869ffee6be9bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR