all cis-8,11,14-Eicosatrienoic acid ≥99% (by GC), clear liquid, Ultrapure

Supplier: Enzo Life Sciences

Synonyms: Dihomo-gamma-linolenic Acid, Dihomo-γ-linolenic acid, (8Z,11Z,14Z)-Icosa-8,11,14-trienoic acid, all cis-8,11,14-Icosatrienoic Acid

BML-FA009-0100 BML-FA009-1000
89158-334EA 237 CAD
89158-334 89158-336
all cis-8,11,14-Eicosatrienoic acid ≥99% (by GC), clear liquid, Ultrapure
all cis-8,11,14-Eicosatrienoic acid

Beneficial effects in cardiovascular diseases and inflammation.


  • Formulation: Clear liquid.


Isolated from borage oil.


Soluble in ethanol, DMSO or acetone.

Formula: C₂₀H₃₄O₂
MW: 306.49 g/mol
MDL Number: MFCD00065721
CAS Number: 1783-84-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f9a9c72155f78b6330bdd5f6472c1b75ab3e21e&vid=a98ed0c828f17bcd7fe4480c35b8ba9d654d58d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99% (GLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR