Arachidonic acid ≥99% (by TLC)
Supplier: Enzo Life Sciences
Synonyms:
(5Z,8Z,11Z,14Z)-Icosa-5,8,11,14-tetraenoic acid
Precursor to a large family of eicosanoids. Acts as a second messenger independent of metabolism. Retrograde messenger in long-term potentiation in the nervous system. Inhibits ras-GAP. Activator for several protein kinase C (PKC) isotypes.
- Eicosanoid precursor and second messenger
Colorless oil.
100mg/ml soluble in DMSO or dimethyl formamide; soluble in 100% ethanol (50mg/ml); sparingly soluble in basic aqueous buffers (1.7mg/ml). To prevent oxidation, the solvent should be purged with an inert gas.
Formula:
C₂₀H₃₂O₂ Storage Temperature: Freezer |
MDL Number:
MFCD00004417 CAS Number: 506-32-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...