SMER28 (synthetic) ≥98% (by TLC)
Supplier: Enzo Life Sciences
|
Danger
|
Small molecule inducer of autophagy important for neurodegenerative disease research.
- Formulation: Off-white solid.
Chemical inducer of autophagy which acts via an mTOR-independent mechanism. It increased autophagosome synthesis and enhanced the clearance of model autophagy substrates such as A53T alpha-synuclein and mutant huntingtin fragments. It reduced Aβ peptide and AAP-CTF levels in an alzheimers model and attenuated mutant huntingtin-fragment toxicity in Huntington's disease models, suggesting therapeutic potential.
Soluble in DMSO (8 mg/mL) or 100% ethanol (2 mg/mL).
Formula:
C₁₁H₁₀BrN₃ Storage Temperature: Freezer |
MDL Number:
MFCD02166825 CAS Number: 307538-42-7 |
|
Specification Test Results
Source | Synthetic |
Purity (TLC) | ≥98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...