Ko143 ≥96% (by HPLC)

Supplier: Enzo Life Sciences

BML-EI396-0005 BML-EI396-0025
89158-270EA 298.62 CAD
89158-270 89158-272
Ko143 ≥96% (by HPLC)
Ko143

Analog of the fungal toxin fumitremorgin C. Potent and selective inhibitor of the breast cancer resistance protein multidrug transporter (BCRP, EC90=26nM). Highly effective for increasing the intracellular drug accumulation and reversing BCRP-mediated multidrug resistance yet has little activity against P-glycoprotein, multidrug resistance-associated protein (MRP1) and other known drug transporters. In mouse models Ko143 was able to restore the intracellular accumulation and abolish the directionality in net flux of dasatinib. It is a highly useful tool for the identification of drug transport processes in complex experimental systems.


  • BCRP inhibitor


White solid.


Soluble in DMSO (>25mg/ml) or 100% ethanol (>25mg/ml).

Formula: C₂₆H₃₅N₃O₅
Storage Temperature: Freezer
CAS Number: 461054-93-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b718f5b1073a8bfa8957b26182d16bce6a68ceb&vid=8bcd8762d67fba000dd8c987da82c6750de8dcd8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR