JZL-184 ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-EI391-0010 BML-EI391-0050
89158-254EA 146.94 CAD
89158-254 89158-256
JZL-184 ≥98% (by TLC)
JZL-184

Potent, selective and irreversible inhibitor of MAG lipase (IC50=8nM vs 4µM for FAAH). JZL-184-treated mice displayed a number of CB1-dependent behavioral effects including analgesia, hypothermia and hypomotility as well as increased levels of 2-arachidonoylglycerol in the brain ( approx. 8-fold, with no effect on anandamide levels). JZL-184 produces a rapid and sustained blockade of brain 2-AG hydrolase activity in mice. It blocks neuropathic pain and enhances retrograde endocannabinoid signaling.


  • Monoacylglycerol lipase inhibitor


Off-white solid.


Soluble in DMSO (10mg/ml).

Formula: C₂₇H₂₄N₂O₉
Storage Temperature: Ambient
MDL Number: MFCD11976911
CAS Number: 1101854-58-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4cf78de472800f3e2cff971c5fed7a39f1dfe43&vid=b5a7483137a4f06fb7639d215b8ffe83c55462e6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR